Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRN Antibody (HL1729) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294859 Supplier Novus Biologicals Supplier No. NBP325758
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

WRN Monoclonal antibody specifically detects WRN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen WRN
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1729
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WRN. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7486
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.