Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRN Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291963 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291963 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291963 Supplier Novus Biologicals Supplier No. NB100472PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WRN Polyclonal specifically detects WRN in Human, Mouse samples. It is validated for Gene Knock-Out, Immunoprecipitation, Western Blot.

Specifications

Antigen WRN
Applications Gene Knock-Out, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation, Knockout Validated
Formulation PBS
Gene Alias DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like
Host Species Rabbit
Immunogen Maps to a region between residues 1400 and the C-terminus (residue 1432) of human Werner Syndrome Helicase using the numbering given in SwissProt entry Q14191 (GeneID 7486).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7486
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.