Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRN Antibody, Novus Biologicals™
SDP

Catalog No. NB100471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100471 Supplier Novus Biologicals Supplier No. NB100471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WRN Polyclonal specifically detects WRN in Human samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen WRN
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:25000, Simple Western reported by internal validation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 2 - 5 ug/mg lysate, Chromatin Immunoprecipitation (ChIP)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q14191
Gene Alias DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like
Gene Symbols WRN
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human Werner Syndrome Helicase using the numbering given in SwissProt entry Q14191 (GeneID 7486).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7486
Test Specificity Reactivity with mouse WRN is unlikely owing to divergence of sequence between this ortholog and human WRN at the site to which the epitope maps.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.