Learn More
Description
Specifications
Specifications
| Antigen | WSB1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp564B0482, SOCS box-containing WD protein SWiP-1, SWIP1DKFZp564A122, WD repeat and SOCS box containing 1, WD repeat and SOCS box-containing 1, WD repeat and SOCS box-containing protein 1, WSB-1 |
| Host Species | Rabbit |
| Immunogen | WSB1 antibody was raised against a 16 amino acid peptide near the amino terminus of human WSB1. The immunogen is located within amino acids 70 - 120 of WSB1. Amino Acid Squence: LPRQNSDGGQKNKPREH |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
