Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WSB1 Antibody (K5P2H10*E10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP250365G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250365G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250365G Supplier Novus Biologicals Supplier No. NBP250365G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WSB1 Monoclonal specifically detects WSB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen WSB1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone K5P2H10*E10
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp564B0482, SOCS box-containing WD protein SWiP-1, SWIP1DKFZp564A122, WD repeat and SOCS box containing 1, WD repeat and SOCS box-containing 1, WD repeat and SOCS box-containing protein 1, WSB-1
Gene Symbols WSB1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide RRVMPTQEVQE
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26118
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.