Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WSB1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 116661AP150UL

Description
WSB1 (WD repeat and SOCS box-containing 1), also known as SWIP1, is a 421 amino acid protein that contains one SOCS box domain and six WD repeats. Expressed as multiple alternatively spliced isoforms, WSB1 is thought to function as part of an SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex that mediates the ubiquitination and proteasomal degradation of target proteins, such as DIO2. Overexpression of WSB1 is implicated in pancreatic cancer progression, suggesting a role for WSB1 in carcinogenesis.Specifications
WSB1 | |
Polyclonal | |
Unconjugated | |
WSB1 | |
SWIP1, WSB 1, WSB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26118 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y6I7 | |
WSB1 | |
WSB1 Fusion Protein Ag2211 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction