Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Antibody (960522), Alexa Fluor 594, Novus Biologicals™
SDP

Catalog No. MB5729AF594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB5729AF594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB5729AF594 Supplier Novus Biologicals Supplier No. MAB5729AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen WT1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 960522
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
Gene Symbols WT1
Host Species Mouse
Immunogen E. coli-derived recombinant human WT1, Met127-Gly249, Accession No. P19544
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7490
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.