Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Antibody (960525), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB347257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347257 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347257 Supplier Novus Biologicals Supplier No. FAB57291MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WT1 Monoclonal antibody specifically detects WT1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen WT1
Applications Flow Cytometry
Classification Monoclonal
Clone 960525
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
Host Species Mouse
Immunogen E.coli-derived recombinant human WT1, Val195-Thr317, Accession # P19544
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7490
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.