Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Antibody (WT1/3477R), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB378112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378112 Supplier Novus Biologicals Supplier No. NBP308966G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

WT1 Monoclonal antibody specifically detects WT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen WT1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone WT1/3477R
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
Host Species Rabbit
Immunogen Recombinant fragment (around aa1-100) of human WT1 protein (exact sequence is proprietary) (Uniprot: P19544 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7490
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.