Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WWOX Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB329876 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329876 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329876 Supplier Novus Biologicals Supplier No. NBP297814PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WWOX Polyclonal antibody specifically detects WWOX in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen WWOX
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias D16S432E, EC 1.1.1, EC 1.1.1.-, FORHHCMA56, FRA16D, Fragile site FRA16D oxidoreductase, SDR41C1, short chain dehydrogenase/reductase family 41C, member 1, WOX1PRO0128, WW domain containing oxidoreductase, WW domain-containing oxidoreductase, WW domain-containing protein WWOX
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human WWOX fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51741
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.