Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WWP1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB048071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048071 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048071 Supplier Novus Biologicals Supplier No. NBP297815DL594

Rabbit Polyclonal Antibody

WWP1 Polyclonal specifically detects WWP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen WWP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AIP5Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 5, Atrophin-1-interacting protein 5, DKFZP434D2111, EC 6.3.2, EC 6.3.2.-, hSDRP1, NEDD4-like E3 ubiquitin-protein ligase WWP1, TGIF-interacting ubiquitin ligase 1, TIUL1, WW domain containing E3 ubiquitin protein ligase 1, WW domain-containing protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human WWP1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11059
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.