Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WWP2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB090934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090934 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090934 Supplier Novus Biologicals Supplier No. NBP300072AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WWP2 Polyclonal specifically detects WWP2 in Human samples. It is validated for ELISA.

Specifications

Antigen WWP2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant WWP2 (O00308; Met1-Glu870).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11060
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.