Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WZ 4003
GREENER_CHOICE

Catalog No. 517710
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
51-771-0 10 mg
51-775-0 50 mg
2 options
This item is not returnable. View return policy

Catalog No. 51-771-0

Supplier: Tocris Bioscience™ 5177/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NUAK1/2 inhibitor

WZ 4003 is a potent and selective NUAK1/2 inhibitor (IC50 values are 20 and 100 nM respectively). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells.

Specifications

Quantity 10 mg
Product Type WZ 4003
Purity >98%
Molecular Weight (g/mol) 496.99
Inhibitors WZ 4003

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.