Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ WZB 117

Click to view available options
Quantity:
5 mg
25 mg
Description
WZB 117 is a GLUT inhibitor (Ki(app) values are 0.2, 10 and 10 μM for GLUT4, GLUT1 and GLUT3, respectively). Competitive inhibitor of net glucose uptake but a noncompetitive inhibitor of sugar efflux from cells. Inhibits passive sugar transport in human erythrocytes and cancer cell lines. Attenuates erythrocyte 3-O-metylglucose (3MG) uptake in vitro (Ki = 6 μM). Suppresses tumor growth in a nude mouse model.
Specifications
Specifications
Chemical Name or Material | 3-Fluoro-1,2-phenylene bis(3-hydroxybenzoate) |
CAS | 1223397-11-2 |
Assay Percent Range | >98% |
Quantity | 25 mg |
Molecular Formula | C20H13FO6 |
Formula Weight | Observed MW: 368.31 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction