Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAB1 Antibody (GPN1/2350), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB009217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009217 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009217 Supplier Novus Biologicals Supplier No. NBP275783R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XAB1 Monoclonal specifically detects XAB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen XAB1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GPN1/2350
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATP(GTP)-binding protein, ATPBD1A, GPN-loop GTPase 1, GTPase, MBD2-interacting protein, MBDin, MBDINXPA binding protein 1, XAB1FLJ51176, XPA-binding protein 1
Gene Symbols GPN1
Host Species Mouse
Immunogen Recombinant full-length human XAB1 protein (Uniprot: Q9HN4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Test Specificity GPN1 is involved in protein synthesis events. It is expressed ubiquitously with highest expression in testis. It binds to the RNA polymerase II- (Poll II) associated proteins RPAP1-3 and to XPA (a protein involved in DNA repair mechanisms), thereby forming an interface with Poll II. Via this interaction, GPN1 is thought to mediate the involvement of Pol II in both protein complex formation and protein chaperone/ scaffolding activities. In addition, GPN1 interacts with components of the integrator and molecular chaperone complexes, further implicating GPN1 in protein assembly. GPN1 contains a cluster of acidic amino acids in its C-terminal region and a series of sequences similar to those found in GTP-binding proteins in its N-terminal region, suggesting that GPN1 has possible GTPase activity.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.