Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAGE1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100262SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100262SS 0.025 mL
NB100262 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100262SS Supplier Novus Biologicals Supplier No. NB100262SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XAGE1 Polyclonal specifically detects XAGE1 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen XAGE1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunoprecipitation 1:10-1:500
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. Q9HD64
Gene Alias CT12.1, CTP9, GAGED2, member 1c, protein XAGE-1, X antigen family, member 1C
Gene Symbols XAGE1B
Host Species Rabbit
Immunogen Synthetic peptide made to an internal sequence within the human XAGE1 protein. [UniProt# Q9HD64]
Molecular Weight of Antigen 20 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 653067
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.