Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XAGE1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | XAGE1 |
---|---|
Dilution | Western Blot 1:500, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100262SS
![]() |
Novus Biologicals
NB100262SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100262
![]() |
Novus Biologicals
NB100262 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
XAGE1 Polyclonal specifically detects XAGE1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
XAGE1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cellular Markers, Signal Transduction, Tumor Suppressors | |
Whole antisera with 0.1% Sodium Azide | |
CT12.1, CTP9, GAGED2, member 1c, protein XAGE-1, X antigen family, member 1C | |
XAGE1B | |
IgG | |
Unpurified | |
20 kDa |
Western Blot 1:500, Immunoprecipitation 1:10-1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q9HD64 | |
653067 | |
Synthetic peptide made to an internal sequence within the human XAGE1 protein. [UniProt# Q9HD64] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title