Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Xamoterol hemifumarate
GREENER_CHOICE

Catalog No. 095010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-501-0 10 mg
09-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-501-0 Supplier Tocris Bioscience™ Supplier No. 0950/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective β1 partial agonist

β1-adrenoceptor-selective partial agonist (pA2 values are 7.4 - 7.8 and 5.2 - 6.2 at β1- and β2-adrenoceptors respectively).

Chemical Identifiers

CAS 73210-73-8
Molecular Formula C36H54N6O14

Specifications

Quantity 10 mg
Formula Weight 397.43
Purity >97%
Molecular Weight (g/mol) 794.856
Chemical Name or Material Xamoterol hemifumarate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.