Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ XAP 044

Click to view available options
Quantity:
10 mg
50 mg
Description
XAP 044 is a potent and selective mGlu7 antagonist (IC50 = 88 nM); inhibits lateral amygdala long term potentiation (LTP) in brain slices from wild type mice. Displays no effects on the LTP of mGlu7 deficient mice. Exhibits novel activity by binding to the mGlu7 Venus flytrap domain (VFTD). Exhibits anti-anxiety effects in rodent models. Brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | XAP 044 |
| Purity | >98% |
| Molecular Weight (g/mol) | 380.13 |
| Inhibitors | XAP 044 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction