Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XBP1 Antibody (525904), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347920 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347920 Supplier Novus Biologicals Supplier No. FAB4257MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XBP1 Monoclonal specifically detects XBP1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen XBP1
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 525904
Conjugate mFluor Violet 610 SE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5
Host Species Mouse
Immunogen E. coli-derived recombinant human XBP1S, Ser188-Leu287, Accession # NP_001073007
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 7494
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.