Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XCL1/Lymphotactin Antibody (MM0604-4G24), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB361502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361502 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361502 Supplier Novus Biologicals Supplier No. NBP212004APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XCL1/Lymphotactin Monoclonal antibody specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot

Specifications

Antigen XCL1/Lymphotactin
Applications Western Blot
Classification Monoclonal
Clone MM0604-4G24
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin)
Host Species Mouse
Immunogen Human recombinant XCL1/Lymphotactin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 6375
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.