Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XCL1/Lymphotactin Antibody (MM0604-4G24) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$170.97 - $554.59
Specifications
| Antigen | XCL1/Lymphotactin |
|---|---|
| Clone | MM0604-4G24 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot.Specifications
| XCL1/Lymphotactin | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| XCL1 | |
| Protein G purified |
| MM0604-4G24 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chemokine Research, Immunology, Innate Immunity | |
| P47992 | |
| 6375 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title