Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XCL2 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB133218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133218 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133218 Supplier Novus Biologicals Supplier No. NBP306178IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XCL2 Polyclonal specifically detects XCL2 in Human samples. It is validated for ELISA.

Specifications

Antigen XCL2
Applications ELISA
Classification Polyclonal
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias C motif chemokine 2, chemokine (C motif) ligand 2, cytokine SCM-1 beta, SCM-1b, SCM1B, SCYC2XC chemokine ligand 2, small inducible cytokine subfamily C, member 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human XCL2 (Q9UBD3; Met1-Gly114).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6846
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.