Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XCR1/CCXCR1 Antibody (1097A), HRP, Novus Biologicals™
SDP

Catalog No. NB346021 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346021 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346021 Supplier Novus Biologicals Supplier No. FAB8571H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

XCR1/CCXCR1 Monoclonal specifically detects XCR1/CCXCR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen XCR1/CCXCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 1097A
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor
Host Species Rabbit
Immunogen E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2829
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.