Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XCT 790
GREENER_CHOICE

Catalog No. 392810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-281-0 10 mg
39-285-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-281-0 Supplier Tocris Bioscience™ Supplier No. 3928/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective ERRα inverse agonist

Selective ERRα inverse agonist (IC50 = ∽400 nM). Displays no antagonist activity at ERRγ or ERα at concentrations below 10 μM. Interferes with PPARγ coactivator-1α (PGC-1α)/ERRα-dependent signaling: inhibits PGC-1α induction of ERRα and MAO-B gene expression and downregulates the constitutive transcriptional activity of ERRα in numerous cell lines.

Chemical Identifiers

CAS 725247-18-7
Molecular Formula C23H13F9N4O3S

Specifications

Quantity 10 mg
Formula Weight 596.42
Purity >99%
Molecular Weight (g/mol) 596.428
Chemical Name or Material XCT 790
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.