Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

xCT Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB300317W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300317W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300317W Supplier Novus Biologicals Supplier No. NB300317AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

xCT Polyclonal specifically detects xCT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen xCT
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, cystine/glutamate transporter, Member 11 (Slc7a11), Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCTCCBR1
Gene Symbols SLC7A11
Host Species Rabbit
Immunogen This xCT Antibody was prepared from a synthetic peptide made to a region within the N-terminus of the mouse xCT protein (between residues 1-50). [Uniprot: Q9WTR6]
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23657
Test Specificity Human. Predicted to react with mouse and canine based on 100% sequence homology. Immunogen sequence has 94% homology to rat.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.