Learn More
Description
Specifications
Specifications
| Antigen | xCT |
| Applications | Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-2 ug/ml, Simple Western, Flow Cytometry 1 - 5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (PMID 21639880), Immunoblotting reported in scientific literature (PMID 28185919) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Alias | Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, cystine/glutamate transporter, Member 11 (Slc7a11), Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCTCCBR1 |
| Gene Symbols | SLC7A11 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
