Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XE 991 dihydrochloride
GREENER_CHOICE

Catalog No. p-4784818
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective KV7 (KCNQ) channel blocker; blocks M-currents

Potent and selective blocker of KV7 (KCNQ) voltage-gated potassium channels. Blocks KV7.2+7.3 (KCNQ2+3)/ M-currents (IC50 = 0.6 - 0.98 μM) and KV7.1 (KCNQ1) homomeric channels (IC50 = 0.75 μM) but is less potent against KV7.1/minK channels (IC50 = 11.1 μM). Augments hippocampal ACh release and is a cognitive enhancer following oral administration in vivo.

Chemical Identifiers

CAS 122955-13-9
Molecular Formula C26H22Cl2N2O

Specifications

Quantity 10 mg
Formula Weight 449.37
Purity >99%
Molecular Weight (g/mol) 449.375
Chemical Name or Material XE 991 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.