Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XIRP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151844 25 μg
NB151843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151844 Supplier Novus Biologicals Supplier No. NBP31195325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XIRP1 Polyclonal antibody specifically detects XIRP1 in Mouse samples. It is validated for Western Blot

Specifications

Antigen XIRP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol.
Gene Alias cardiomyopathy associated 1, CMYA1, DKFZp779C1255, xin actin-binding repeat containing 1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser295 conjugated to KLH. (UniProt: O70373)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165904
Target Species Mouse
Content And Storage Store at -20C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.