Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XIRP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | XIRP1 |
---|---|
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
XIRP1 Polyclonal antibody specifically detects XIRP1 in Mouse samples. It is validated for Western BlotSpecifications
XIRP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
cardiomyopathy associated 1, CMYA1, DKFZp779C1255, xin actin-binding repeat containing 1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser295 conjugated to KLH. (UniProt: O70373) | |
Antigen Affinity-purified |
Western Blot 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. | |
165904 | |
Primary | |
Store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title