Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ XL 184

Click to view available options
Quantity:
10 mg
50 mg
Description
XL 184 is a potent VEGFR inhibitor (IC50 = 0.035 nM); also inhibits c-Met, KIT, RET, FLT4, AXL, FLT3, FLT1 and Tie2 (IC50 values are 1.3, 4.6, 5.2, 6, 7, 11.3, 12 and 14.3 nM, respectively). Induces intratumoral hypoxia and apoptosis. Reduces tumor invasion and metastasis in vivo. Antiangiogenic.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | XL 184 |
| Purity | >98% |
| Molecular Weight (g/mol) | 501.51 |
| Inhibitors | XL 184 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction