Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XLF Antibody (OTI1B5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274880A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274880A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274880A30 Supplier Novus Biologicals Supplier No. NBP274880AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XLF Monoclonal specifically detects XLF in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen XLF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B5
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor
Gene Symbols NHEJ1
Host Species Mouse
Immunogen Full length recombinant protein of human XLF (NP_079058) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 79840.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.