Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XMD 8-92
GSA_VAGREENER_CHOICESDP

Catalog No. 413210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-3210 10 mg
41-325-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-3210 Supplier Tocris Bioscience™ Supplier No. 4132/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

ERK5/BMK1 inhibitor; also BRD4 inhibitor

ERK5 (BMK1) and BRD4 inhibitor (Kd values are 80 and 190 nM, respectively). Also inhibits DCAMKL2, PLK4 and TNK1 (Kd values are 190, 600 and 890 nM). Blocks growth factor-induced activation of cellular BMK1 and reduces BMK1 activity in in vitro kinase assays. Also reduces BMK1-dependent transactivating activity of MEF2C. Inhibits proliferation in a variety of cancer cell lines; blocks tumor cell proliferation and tumor-associated angiogenesis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1234480-50-2
Molecular Formula C26H30N6O3

Specifications

Quantity 10 mg
Formula Weight 474.55
Purity >98%
Molecular Weight (g/mol) 474.565
Chemical Name or Material XMD 8-92
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.