Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPB Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB176459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176459 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176459 Supplier Novus Biologicals Supplier No. NB10061060B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XPB Polyclonal specifically detects XPB in Human, Mouse, Rabbit, Virus samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.

Specifications

Antigen XPB
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Basic transcription factor 2 89 kDa subunit, BTF2, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 3 (xeroderma pigmentosum group B complementing), GTF2H, RAD25, TFIIH, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, Xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B, XPBC, XPBTFIIH 89 kDa subunit
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 732 and 782 of human excision repair cross-complementing rodent repair deficiency, complementation group 3 using the numbering given in entry NP_000113.1 (GeneID 2071).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2071
Target Species Human, Mouse, Rabbit, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.