Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPD Antibody, Novus Biologicals™
SDP

Catalog No. NB396855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396855 25 μL
NBP238382 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396855 Supplier Novus Biologicals Supplier No. NBP23838225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XPD Polyclonal specifically detects XPD in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen XPD
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P18074
Gene Alias Basic transcription factor 2 80 kDa subunit, BTF2 p80, COFS2, CXPD, DNA excision repair protein ERCC-2, DNA repair protein complementing XP-D cells, EC 3.6.1, EC 3.6.4.12, EM9TFIIH basal transcription factor complex 80 kDa subunit, excision repair cross-complementing rodent repair deficiency, complementationgroup 2, MAG, MGC102762, MGC126218, MGC126219, TFIIH 80 kDa subunit, TFIIH basal transcription factor complex helicase subunit, TFIIH basal transcription factor complex helicase XPD subunit, TTD, xeroderma pigmentosum complementary group D, Xeroderma pigmentosum group D-complementing protein, XPDC, XPDTFIIH p80
Gene Symbols ERCC2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AHNIDNVCIDSMSVNLTRRTLDRCQGNLETLQKTVLRIKETDEQRLRDEYRRLVEGLREASAARETDAHLANPVLPDEVLQEAVPGSIRTAEHFLGFL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2068
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.