Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPE/DDB2 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB056693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056693 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056693 Supplier Novus Biologicals Supplier No. NBP298940H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XPE/DDB2 Polyclonal specifically detects XPE/DDB2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen XPE/DDB2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human XPE/DDB2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 1643
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.