Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Antibody (OTI4E11), PerCP, Novus Biologicals™
SDP

Catalog No. NB274883PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274883PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274883PCP Supplier Novus Biologicals Supplier No. NBP274883PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XPF Monoclonal specifically detects XPF in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen XPF
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4E11
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster
Gene Symbols ERCC4
Host Species Mouse
Immunogen Full length recombinant protein of human XPF (NP_005227) produced in HEK293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2072.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.