Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPG Antibody (8H7/XPG), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250366AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250366AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250366AF Supplier Novus Biologicals Supplier No. NBP250366AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XPG Monoclonal specifically detects XPG in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.

Specifications

Antigen XPG
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 8H7/XPG
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR
Gene Symbols ERCC5
Host Species Mouse
Immunogen Recombinant human XPG protein produced in baculovirus
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2073
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.