Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPG Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB050456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050456 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050456 Supplier Novus Biologicals Supplier No. NBP297984R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XPG Polyclonal specifically detects XPG in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen XPG
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human XPG fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 2073
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.