Learn More
Eppendorf™ Xplorer™ 12-channel plus epT.I.P.S. Bundle
Catalog No. p-7112272 Shop All Eppendorf North America Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
15 to 300 μL
5 to 100 μL
Color:
Gray
Orange
Yellow
Eppendorf Xplorer 12-channel pipette is designed to aspirate and dispense liquid volumes from 0.5μL to 10 mL while eliminating many of potential error risks associated with manual pipetting.
  • Intuitive operating concept for quick and easy work
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle
  • Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 mL and 10 mL)
  • Adjustment to external conditions (geographic height, liquids with high density, or similar factors)
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Rechargeable
Compatible Tips epT.I.P.S
Color Gray
Number of Channels 12
Product Line Xplorer
Type Electronic Pipette Bundle
Volume (Metric) 0.5 to 10 μL
Display Digital
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.