Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPNPEP3 Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA503339

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-4259


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The function of this protein remains unknown.
Specifications

Specifications

XPNPEP3
Monoclonal
0.86 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
B5DEQ3, B7ZMP1, Q9NQH7
Xpnpep3
Full length human recombit protein of human XPNPEP3 produced in HEK293 cell.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4B8
Unconjugated
XPNPEP3
APP3, ICP55, NPHPL1
Mouse
Affinity Chromatography
RUO
321003, 481237, 63929, 685823
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.