Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPNPEP3 Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene
SDP

Catalog No. p-200012835
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0971 30 μL
50-167-4259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0971 Supplier OriGene Supplier No. TA503339S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XPNPEP3 Monoclonal antibody specifically detects XPNPEP3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen XPNPEP3
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B8
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Irf6
Gene Accession No. O14896
Gene Alias Aminopeptidase P3; APP3; E430012M05Rik; ICP55; Intermediate Cleaving Peptidase 55; LOW QUALITY PROTEIN: xaa-Pro aminopeptidase 3; NPHPL1; probable Xaa-Pro aminopeptidase 3; putative Xaa-Pro aminopeptidase 3; Xaa-Pro aminopeptidase 3; xaa-Pro aminopeptidase 3; probable Xaa-Pro aminopeptidase 3; XPNPEP3; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3; X-prolyl aminopeptidase 3, mitochondrial; zgc:76905; zgc:76905 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human IRF6 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3664
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.