Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPO6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220925 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220925 Supplier Novus Biologicals Supplier No. NBP220925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XPO6 Polyclonal specifically detects XPO6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen XPO6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96QU8
Gene Alias Exp6, exportin 6, exportin-6, FLJ22519, KIAA0370EXP6, RAN binding protein 20, Ran-binding protein 20, RANBP20
Gene Symbols XPO6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human XPO6. The exact sequence is proprietary.
Molecular Weight of Antigen 129 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23214
Test Specificity Human. Species reactivity based on sequence identity: Mouse (97%), Rat (97%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.