Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPV/DNA polymerase eta Antibody, Novus Biologicals™
SDP

Catalog No. NB10060423 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060423 Supplier Novus Biologicals Supplier No. NB10060423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XPV/DNA polymerase eta Polyclonal specifically detects XPV/DNA polymerase eta in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen XPV/DNA polymerase eta
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_006493.1
Gene Alias DNA polymerase eta, EC 2.7.7.7, polymerase (DNA directed), eta, RAD30, RAD30 homolog A, RAD30AFLJ16395, Xeroderma pigmentosum variant type protein, XP-V, XPVFLJ21978
Gene Symbols POLH
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 640 and 690 of human polymerase (DNA directed), eta using the numbering given in entry NP_006493.1 (GeneID 5429).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5429
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.