Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC3 Antibody (10F1/6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB383304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383304 Supplier Novus Biologicals Supplier No. NBP308967AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XRCC3 Monoclonal antibody specifically detects XRCC3 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen XRCC3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10F1/6
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3
Host Species Mouse
Immunogen Recombinant full-length human XRCC3 protein (Uniprot: O43542)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7517
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.