Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB10165A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10165A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10165A532 Supplier Novus Biologicals Supplier No. NB100165AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

XRCC3 Polyclonal specifically detects XRCC3 in Drosophila, Hamster, Human samples. It is validated for ICC/IF (Negative), Immunoprecipitation, Western Blot.

Specifications

Antigen XRCC3
Applications ICC/IF (Negative), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3
Gene Symbols XRCC3
Host Species Rabbit
Immunogen A synthetic partial peptide of human XRCC3. [UniProtNo. O43542]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7517.0
Target Species Drosophila, Hamster, Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.