Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC4 Antibody (OTI4H9), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP274896A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274896A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274896A32 Supplier Novus Biologicals Supplier No. NBP274896AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XRCC4 Monoclonal specifically detects XRCC4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen XRCC4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4H9
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias complementing defective, repair in Chinese hamster, X-ray repair complementing defective repair in Chinese hamster cells 4, X-ray repair cross-complementing protein 4
Gene Symbols XRCC4
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human XRCC4 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7518.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.