Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XTP3TPA Antibody (03), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB351112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351112 Supplier Novus Biologicals Supplier No. NBP306606MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XTP3TPA Monoclonal antibody specifically detects XTP3TPA in Human samples. It is validated for ELISA

Specifications

Antigen XTP3TPA
Applications ELISA
Classification Monoclonal
Clone 03
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human XTP3TPA (Accession#: NP_077001.1; Met1-Thr170).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 79077
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.