Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XTP3TPA Antibody (074), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB071961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB071961 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB071961 Supplier Novus Biologicals Supplier No. NBP290294R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

XTP3TPA Monoclonal specifically detects XTP3TPA in Human samples. It is validated for ELISA.

Specifications

Antigen XTP3TPA
Applications ELISA
Classification Monoclonal
Clone 74
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human XTP3TPA (NP_077001.1; Met1-Thr170).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 79077
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.