Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XTP4 Antibody (OTI1E8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP274899A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274899A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274899A70 Supplier Novus Biologicals Supplier No. NBP274899AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

XTP4 Monoclonal specifically detects XTP4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen XTP4
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1E8
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C35, chromosome 17 open reading frame 37, HBV XAg-transactivated protein 4, HBV X-transactivated gene 4 protein, MGC14832, ORB3, protein C17orf37, Protein C35, XTP4RDX12
Gene Symbols MIEN1
Host Species Mouse
Immunogen Full length recombinant protein of human XTP4 (NP_115715) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84299.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.